siRNA Details
Gene Symbol: Acads | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007383.3 | NP_031409.2 | 2 | 196 | ||
GenBank mRNA | AK155361.1 | - | 2 | 175 | ||
AK169428.1 | - | 2 | 133 | |||
BC016259.1 | - | 2 | 163 | 4913813 | ||
BC055063.1 | - | 2 | 137 | |||
BY077302.1 | - | 2 | 197 | |||
L11163.1 | - | 2 | 120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
