siRNA Details
Gene Symbol: Acads | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007383.3 | NP_031409.2 | 10 | 1241 | ||
GenBank mRNA | AK155361.1 | - | 10 | 1220 | ||
AK169428.1 | - | 10 | 1178 | |||
BC016259.1 | - | 10 | 1208 | 4913813 | ||
BC055063.1 | - | 9 | 1394 | |||
L11163.1 | - | 10 | 1165 |

More Information
Other Species Targeted

Gene Ontology Categories:
