siRNA Details
Gene Symbol: Hsd17b11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053262.3 | NP_444492.1 | 3 | 441 | ||
GenBank mRNA | AF304306.1 | - | 3 | 437 | ||
AK049355.1 | - | 3 | 485 | |||
AK154803.1 | - | 3 | 456 | |||
AK155174.1 | - | 3 | 456 | |||
AK155202.1 | - | 3 | 461 | |||
AK155327.1 | - | 3 | 461 | |||
AK158327.1 | - | 3 | 456 | |||
AK170939.1 | - | 3 | 456 | |||
AY053570.1 | - | 3 | 455 | |||
BC038340.1 | - | 3 | 441 | 4458725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
