siRNA Details
Gene Symbol: Crisp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009639.2 | NP_033769.1 | 105 | |||
GenBank mRNA | BC022573.1 | - | 2, 3 | 126 | 4925349 | |
BC114345.2 | - | 106 | ||||
BC132536.1 | - | 104 | 40130813 | |||
BC132538.1 | - | 104 | 40130815 | |||
L05560.1 | - | 105 | ||||
M92850.1 | - | 2, 3 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
