siRNA Details
Gene Symbol: Agtrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301281.1 | NP_001288210.1 | 5 | 723 | ||
NM_009642.4 | NP_033772.2 | 5 | 770 | |||
GenBank mRNA | AK007383.1 | - | 5 | 707 | ||
AK007502.1 | - | 5 | 682 | |||
AK036598.1 | - | 5 | 699 | |||
AK152467.1 | - | 5 | 770 | |||
BC046820.1 | - | 5 | 756 | 6400135 | ||
BC057196.1 | - | 5 | 675 | 5249164 |

More Information
Other Species Targeted

Gene Ontology Categories:
