siRNA Details
Gene Symbol: Alox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009662.2 | NP_033792.1 | 3 | 549 | ||
GenBank mRNA | AK089344.1 | - | 3 | 503 | ||
AK136868.1 | - | 3 | 517 | |||
AK137481.1 | - | 3 | 507 | |||
AK169938.1 | - | 3 | 506 | |||
AK170016.1 | - | 3 | 406 | |||
AK170434.1 | - | 3 | 506 | |||
AK170879.1 | - | 3 | 505 | |||
AK171191.1 | - | 3 | 498 | |||
AK171413.1 | - | 3 | 506 | |||
BC139102.1 | - | 3 | 476 | 8862124 | ||
BC141213.1 | - | 3 | 476 | 9055825 | ||
L42198.1 | - | 3 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
