siRNA Details
Gene Symbol: Amd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007444.3 | NP_031470.3 | 1 | 666 |
Gene Symbol: Amd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009665.5 | NP_033795.1 | 3, 4 | 664 | ||
GenBank mRNA | AK146840.1 | - | 3, 4 | 663 | ||
AK166187.1 | - | 3, 4 | 658 | |||
AK168165.1 | - | 3, 4 | 661 | |||
AK171238.1 | - | 3, 4 | 663 | |||
BC011110.1 | - | 3, 4 | 649 | 3672700 | ||
BC071220.1 | - | 3, 4 | 647 | 5012910 | ||
BC080791.1 | - | 3, 4 | 428 | 4014056 | ||
BC092072.1 | - | 3, 4 | 645 | 4505452 | ||
BC138696.1 | - | 3, 4 | 402 | 8861718 | ||
BC138697.1 | - | 3, 4 | 402 | 8861719 | ||
CT010192.1 | - | 3, 4 | 334 | |||
D12780.1 | - | 3, 4 | 660 | |||
Z14986.1 | - | 3, 4 | 591 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031011, BC061532, AY724509, FM031711, FQ234346, and others.
Gene Ontology Categories:
