siRNA Details
Gene Symbol: Ube3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054093.2 | NP_473434.2 | 22 | 2962 | ||
GenBank mRNA | AF244362.3 | - | 21 | 2597 | ||
AK088518.1 | - | 21 | 2658 | |||
BC023956.1 | - | 22 | 2731 | 5321098 | ||
BC026415.1 | - | 4 | 470 | |||
BC034059.1 | - | 16 | 1966 | 4163877 | ||
BC096743.1 | - | 22 | 2985 | 30549276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
