siRNA Details
Gene Symbol: Ank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009670.4 | NP_033800.2 | 18 | 2979 | ||
NM_146005.3 | NP_666117.2 | 41 | 6038 | |||
NM_170687.3 | NP_733788.2 | 18 | 3567 | |||
NM_170688.2 | NP_733789.1 | 41 | 5450 | |||
NM_170689.2 | NP_733790.2 | 40 | 5984 | |||
NM_170690.2 | NP_733791.2 | 40 | 5975 | |||
NM_170728.2 | NP_733924.2 | 39 | 5333 | |||
NM_170729.2 | NP_733925.2 | 39 | 5921 | |||
NM_170730.2 | NP_733926.2 | 16 | 2669 | |||
GenBank mRNA | AF212924.1 | - | 16 | 2169 | ||
AK033389.1 | - | 3 | 865 | |||
AK035681.1 | - | 18 | 3295 | |||
BC021657.1 | - | 39 | 5203 | 4188590 | ||
BC026152.1 | - | 18 | 2581 | |||
BC062122.1 | - | 7 | 3287 | |||
L40631.1 | - | 18 | 3570 | |||
L40632.1 | - | 41 | 5580 | |||
U89274.1 | - | 20 | 2596 | |||
U89275.1 | - | 20 | 3184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
