siRNA Details
Gene Symbol: Aoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009675.2 | NP_033805.1 | 4 | 2391 | ||
GenBank mRNA | AF054831.1 | - | 4 | 2406 | ||
AK040729.1 | - | 1218 | ||||
BC080857.1 | - | 4 | 2237 | 6854465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
