siRNA Details
Gene Symbol: Ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF084575.1 | - | 4, 5 | 423 | ||
AK002565.1 | - | 4, 5 | 449 | |||
AK137489.1 | - | 4, 5 | 444 | |||
AK152100.1 | - | 4, 5 | 455 | |||
AK184535.1 | - | 2, 3 | 103 | |||
AK194047.1 | - | 2, 3 | 205 | |||
BC012656.1 | - | 4, 5 | 434 | 4187607 | ||
BC108327.1 | - | 4, 5 | 442 | 30713247 | ||
BC131985.1 | - | 4, 5 | 424 | 40130262 | ||
BC145234.1 | - | 4, 5 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
