siRNA Details
Gene Symbol: Axin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015732.4 | NP_056547.3 | 10, 11 | 2739 | ||
GenBank mRNA | AF073788.1 | - | 10, 11 | 2604 | ||
AF205889.1 | - | 9, 10 | 2444 | |||
AK049192.1 | - | 10, 11 | 2684 | |||
AK084644.1 | - | 3, 4 | 524 | |||
AK198476.1 | - | 2, 3 | 260 | |||
AK202759.1 | - | 2, 3 | 260 | |||
BC057338.1 | - | 10, 11 | 2722 | 6827741 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
