siRNA Details
Gene Symbol: Cacnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037099.2 | NP_001032176.1 | 5 | 767 | ||
NM_001285426.1 | NP_001272355.1 | 5 | 551 | |||
NM_001285427.1 | NP_001272356.1 | 5 | 847 | |||
NM_001285428.1 | NP_001272357.1 | 4 | 476 | |||
NM_146123.3 | NP_666235.1 | 4 | 687 | |||
GenBank mRNA | AK032311.1 | - | 5 | 991 | ||
AK038633.1 | - | 5 | 803 | |||
AK043850.1 | - | 4 | 630 | |||
AK079616.1 | - | 4 | 637 | |||
AK147338.1 | - | 5 | 741 | |||
AK158606.1 | - | 4 | 478 | |||
BC026479.1 | - | 4 | 473 | 4501980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
