siRNA Details
Gene Symbol: Ddr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198831.1 | NP_001185760.1 | 4 | 358 | ||
NM_001198833.1 | NP_001185762.1 | 4 | 477 | |||
NM_007584.2 | NP_031610.1 | 4 | 533 | |||
NM_172962.1 | NP_766550.1 | 4 | 533 | |||
GenBank mRNA | AF026259.1 | - | 4 | 375 | ||
AK031442.1 | - | 4 | 436 | |||
AK138698.1 | - | 4 | 345 | |||
AK143088.1 | - | 4 | 426 | |||
BC006836.1 | - | 4 | 439 | |||
BC037108.1 | - | 5 | 612 | |||
BC065998.1 | - | 4 | 369 | 5686085 | ||
CK790776.1 | - | 4 | 358 | |||
L57509.1 | - | 4 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
