siRNA Details
Gene Symbol: Cd37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290802.1 | NP_001277731.1 | 7 | 736 | ||
NM_001290804.1 | NP_001277733.1 | 6 | 661 | |||
NM_007645.4 | NP_031671.1 | 6 | 685 | |||
GenBank mRNA | AK154250.1 | - | 6 | 725 | ||
AK154466.1 | - | 7 | 706 | |||
AK155663.1 | - | 6 | 662 | |||
AK155966.1 | - | 6 | 650 | |||
BC019402.1 | - | 6 | 722 | 4013070 | ||
BE849269.1 | - | 6 | 603 | |||
CT010217.1 | - | 6 | 574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
