siRNA Details
Gene Symbol: Ccr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009915.2 | NP_034045.1 | 3 | 1171 | ||
GenBank mRNA | AK039755.1 | - | 1 | 21 | ||
AK046579.1 | - | 1405 | ||||
AK138599.1 | - | 2 | 1484 | |||
AK162705.1 | - | 1 | 177 | |||
BC138803.1 | - | 2 | 1131 | 8861825 | ||
BC138804.1 | - | 2 | 1131 | 8861826 | ||
U47035.1 | - | 2 | 1080 | |||
U51717.1 | - | 2 | 1129 | |||
U56819.1 | - | 3 | 1171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
