siRNA Details
Gene Symbol: Coq7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF053770.1 | - | 1, 2 | 196 | ||
AF080580.1 | - | 2, 3 | 259 | |||
AF098949.1 | - | 2, 3 | 325 | |||
AK158380.1 | - | 2, 3 | 310 | |||
BC038681.1 | - | 2, 3 | 292 | 1380996 | ||
BY284216.1 | - | 2, 3 | 364 | |||
U81277.1 | - | 1, 2 | 41 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
