siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244891.1 | NP_001231820.1 | 12 | 1450 | ||
NM_001244903.1 | NP_001231832.1 | 11 | 1397 | |||
NM_017368.3 | NP_059064.2 | 12 | 1549 | |||
NM_198683.2 | NP_941955.1 | 11 | 1397 | |||
GenBank mRNA | AB050499.1 | - | 8 | 861 | ||
AF267535.1 | - | 10 | 1153 | |||
AF314172.1 | - | 10 | 994 | |||
AJ007987.1 | - | 10 | 1142 | |||
AK014492.1 | - | 11 | 1313 | |||
AK032751.1 | - | 1 | 1941 | |||
AK143698.1 | - | 12 | 1351 | |||
AK156725.1 | - | 11 | 1310 | |||
AK183548.1 | - | 2 | 156 | |||
AK188588.1 | - | 2 | 156 | |||
AK196827.1 | - | 1 | 93 | |||
AK200025.1 | - | 1 | 105 | |||
AK208664.1 | - | 1 | 91 | |||
BC021393.1 | - | 11 | 1262 | |||
CF745326.1 | - | 1 | 87 | |||
X61451.1 | - | 7 | 847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
