siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244891.1 | NP_001231820.1 | 7 | 796 | ||
NM_001244903.1 | NP_001231832.1 | 6 | 743 | |||
NM_017368.3 | NP_059064.2 | 7 | 895 | |||
NM_198683.2 | NP_941955.1 | 6 | 743 | |||
GenBank mRNA | AB050499.1 | - | 3 | 204 | ||
AF267535.1 | - | 5 | 499 | |||
AF314172.1 | - | 5 | 337 | |||
AJ007987.1 | - | 5 | 488 | |||
AK014492.1 | - | 6 | 656 | |||
AK143698.1 | - | 7 | 697 | |||
AK156725.1 | - | 6 | 656 | |||
AK161213.1 | - | 6 | 703 | |||
AK210607.1 | - | 2 | 68 | |||
AK218377.1 | - | 2 | 68 | |||
BC021393.1 | - | 6 | 608 | |||
X61451.1 | - | 2 | 191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
