siRNA Details
Gene Symbol: Cyp2b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009999.4 | NP_034129.1 | 3 | 397 | ||
GenBank mRNA | AF128849.1 | - | 3 | 379 | ||
AF129405.1 | - | 2 | 193 | |||
AI304065.1 | - | 3 | 351 | |||
AK028103.1 | - | 3 | 397 | |||
BC060973.1 | - | 3 | 391 | 30301385 | ||
M21856.1 | - | 3 | 379 | |||
X99715.1 | - | 3 | 375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
