siRNA Details
Gene Symbol: Dgka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016811.2 | NP_058091.2 | 8, 9 | 932 | ||
GenBank mRNA | AF085219.1 | - | 7, 8 | 618 | ||
BC006713.1 | - | 8, 9 | 932 | 3584592 | ||
JN832677.1 | - | 9, 10 | 964 | |||
JN832678.1 | - | 8, 9 | 951 | |||
JN832679.1 | - | 8, 9 | 951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
