siRNA Details
Gene Symbol: Dgka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016811.2 | NP_058091.2 | 18 | 1761 | ||
GenBank mRNA | AF085219.1 | - | 17 | 1447 | ||
AK008063.1 | - | 6 | 425 | |||
AK008314.1 | - | 7 | 432 | |||
BC006713.1 | - | 18 | 1761 | 3584592 | ||
JN832677.1 | - | 18 | 1704 | |||
JN832678.1 | - | 18 | 1809 | |||
JN832679.1 | - | 18 | 1780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
