siRNA Details
Gene Symbol: Dnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301737.1 | NP_001288666.1 | 2 | 344 | ||
NM_010065.3 | NP_034195.2 | 2 | 344 | |||
GenBank mRNA | AK011651.1 | - | 2 | 311 | ||
AK144142.1 | - | 2 | 295 | |||
AK220483.1 | - | 2 | 286 | |||
BC034679.1 | - | 2 | 298 | 4506865 | ||
BC058623.1 | - | 2 | 278 | 5706803 | ||
BY297150.1 | - | 2 | 344 | |||
L31395.1 | - | 2 | 241 | |||
L31397.1 | - | 2 | 297 |

More Information
Other Species Targeted

Gene Ontology Categories:
