siRNA Details
Gene Symbol: Dvl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007888.3 | NP_031914.3 | 8, 9 | 1174 | ||
GenBank mRNA | AK050614.1 | - | 8, 9 | 978 | ||
AK144001.1 | - | 8, 9 | 1205 | |||
AK146822.1 | - | 8, 9 | 1174 | |||
AK159895.1 | - | 8, 9 | 1007 | |||
AK168376.1 | - | 8, 9 | 1082 | |||
BC053050.1 | - | 8, 9 | 1088 | 6402000 | ||
U24160.1 | - | 8, 9 | 1186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
