siRNA Details
Gene Symbol: Efna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007910.2 | NP_031936.2 | 2 | 400 | ||
GenBank mRNA | AK012195.1 | - | 2 | 404 | ||
AK142604.1 | - | 2 | 401 | |||
U90663.1 | - | 2 | 400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
