siRNA Details
Gene Symbol: Efnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007911.5 | NP_031937.1 | 5 | 2286 | ||
GenBank mRNA | AK048305.1 | - | 5 | 2230 | ||
BC052001.1 | - | 5 | 2103 | 5695429 | ||
BC058617.1 | - | 5 | 2219 | 5695418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
