siRNA Details
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007963.2 | NP_031989.2 | 4 | 1079 | ||
GenBank mRNA | AJ001482.1 | - | 2 | 235 | ||
AK033517.1 | - | 4 | 513 | |||
AK051098.1 | - | 4 | 936 | |||
AK085548.1 | - | 4 | 1080 | |||
BC076620.1 | - | 4 | 1031 | |||
BC139762.1 | - | 2 | 197 | 40109903 | ||
JQ665270.1 | - | 2 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
