siRNA Details
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007963.2 | NP_031989.2 | 12 | 2382 | ||
GenBank mRNA | AJ001482.1 | - | 11 | 2537 | ||
AK033517.1 | - | 12 | 1816 | |||
BC076620.1 | - | 12 | 2334 | |||
BC139762.1 | - | 10 | 1500 | 40109903 | ||
JQ665270.1 | - | 10 | 2502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
