siRNA Details
Gene Symbol: Fasn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007988.3 | NP_032014.3 | 36 | 6633 | ||
GenBank mRNA | AF127033.1 | - | 36 | 6150 | ||
AK080374.1 | - | 10 | 1530 | |||
AK147214.1 | - | 36 | 6329 | |||
AK147694.1 | - | 36 | 6397 | |||
AK171499.1 | - | 36 | 6265 | |||
BC010310.1 | - | 5 | 620 | |||
BC046513.1 | - | 36 | 6187 | 4196969 | ||
BC059850.1 | - | 8 | 1165 | |||
U89406.1 | - | 3 | 255 | |||
X13135.1 | - | 8 | 1108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
