siRNA Details
Gene Symbol: Fgf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008002.4 | NP_032028.1 | 1 | 992 | ||
GenBank mRNA | AK034742.1 | - | 1 | 558 | ||
AK045267.1 | - | 1 | 993 | |||
AK085244.1 | - | 1 | 314 | |||
AK141633.1 | - | 1 | 314 | |||
BC048229.1 | - | 1 | 657 | 6313081 | ||
D89080.1 | - | 1 | 307 | |||
U94517.1 | - | 1 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
