siRNA Details
Gene Symbol: G6pc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008061.3 | NP_032087.2 | 5 | 1085 | ||
GenBank mRNA | AK050279.1 | - | 5 | 1100 | ||
AK052656.1 | - | 5 | 1100 | |||
BC013448.1 | - | 5 | 1085 | 4237115 | ||
U00445.1 | - | 5 | 1100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
