siRNA Details
Gene Symbol: Slc6a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133661.3 | NP_598422.2 | 11 | 1545 | ||
GenBank mRNA | BC019211.1 | - | 11 | 1549 | 5053116 | |
M97632.1 | - | 11 | 1285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
