siRNA Details
Gene Symbol: Gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010255.3 | NP_034385.1 | 5 | 602 | ||
GenBank mRNA | AF015887.1 | - | 5 | 535 | ||
AK140688.1 | - | 5 | 651 | |||
AK158087.1 | - | 5 | 545 | |||
AK178068.1 | - | 1 | 21 | |||
AK217844.1 | - | 1 | 21 | |||
BC049233.1 | - | 5 | 588 | 4195030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
