siRNA Details
Gene Symbol: Gata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008090.5 | NP_032116.4 | 6 | 1527 | ||
GenBank mRNA | AB000096.2 | - | 6 | 1419 | ||
AK004551.1 | - | 6 | 1524 | |||
AK004675.1 | - | 6 | 1856 | |||
AK154792.1 | - | 6 | 1525 | |||
AK154981.1 | - | 6 | 1528 | |||
BC107009.2 | - | 6 | 1431 | 40047573 | ||
BC107010.1 | - | 5 | 1204 | 40047574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
