siRNA Details
Gene Symbol: Grpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033473.1 | - | 3 | 1211 | ||
BC113145.1 | - | 3 | 1099 | 40047100 | ||
M57922.1 | - | 3 | 1227 | |||
M61000.1 | - | 3 | 1200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
