siRNA Details
Gene Symbol: Gzmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010372.2 | NP_034502.2 | 3 | 324 | ||
GenBank mRNA | AK131671.1 | - | 3 | 325 | ||
BC141122.1 | - | 3 | 279 | 9055734 | ||
BC145433.1 | - | 279 | 9053991 | |||
BC145434.1 | - | 3 | 279 | |||
J03255.1 | - | 3 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
