siRNA Details
Gene Symbol: H2-DMb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK154923.1 | - | 5, 6 | 881 | ||
BC052864.1 | - | 6, 7 | 1032 | |||
BC141105.1 | - | 5, 6 | 843 | 9055717 | ||
BC145385.1 | - | 5, 6 | 843 | 9053943 | ||
BC145386.1 | - | 5, 6 | 843 | 9053944 | ||
BC171960.1 | - | 5, 6 | 843 | 9054246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
