siRNA Details
Gene Symbol: Hoxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079869.1 | NP_001073338.1 | 4 | 2591 | ||
NM_010458.2 | NP_034588.2 | 2 | 2514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
