siRNA Details
Gene Symbol: Il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080147.1 | - | 11 | 1250 | ||
AK082889.1 | - | 11 | 1244 | |||
AK089984.1 | - | 11 | 1280 | |||
AK143292.1 | - | 11 | 1250 | |||
AK154675.1 | - | 11 | 1305 | |||
BC021472.1 | - | 7 | 703 | |||
BC052425.1 | - | 11 | 1272 | 6835500 | ||
BC059939.1 | - | 11 | 1240 | 6477224 | ||
S80963.1 | - | 11 | 1277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
