siRNA Details
Gene Symbol: Irx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010573.2 | NP_034703.2 | 2 | 1329 | ||
GenBank mRNA | AF165984.1 | - | 2 | 1094 | ||
AK133937.1 | - | 2 | 1329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
