siRNA Details
Gene Symbol: Itgav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008402.3 | NP_032428.2 | 7 | 1068 | ||
GenBank mRNA | AK149984.1 | - | 7 | 970 | ||
BC133682.1 | - | 7 | 779 | |||
U14135.1 | - | 7 | 950 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
