siRNA Details
Gene Symbol: Itgav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008402.3 | NP_032428.2 | 5 | 913 | ||
GenBank mRNA | AK149984.1 | - | 5 | 815 | ||
BC133682.1 | - | 5 | 625 | |||
U14135.1 | - | 5 | 795 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008762000.
Gene Ontology Categories:
