siRNA Details
Gene Symbol: Itgav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008402.3 | NP_032428.2 | 22 | 2557 | ||
GenBank mRNA | AK149984.1 | - | 22 | 2458 | ||
AK154307.1 | - | 10 | 953 | |||
BC133682.1 | - | 22 | 2268 | |||
U14135.1 | - | 22 | 2439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
