siRNA Details
Gene Symbol: Itm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008410.2 | NP_032436.1 | 5 | 755 | ||
GenBank mRNA | AB030203.1 | - | 5 | 775 | ||
AB030204.1 | - | 4 | 571 | |||
AK002599.1 | - | 5 | 718 | |||
AK003224.1 | - | 5 | 722 | |||
AK003587.1 | - | 5 | 744 | |||
AK003928.1 | - | 5 | 743 | |||
AK005125.1 | - | 5 | 746 | |||
AK076139.1 | - | 5 | 764 | |||
AK077482.1 | - | 5 | 757 | |||
AK088270.1 | - | 5 | 742 | |||
AK150378.1 | - | 5 | 775 | |||
AK150396.1 | - | 5 | 774 | |||
AK151714.1 | - | 5 | 774 | |||
AK151889.1 | - | 5 | 775 | |||
AK152068.1 | - | 5 | 763 | |||
AK152516.1 | - | 5 | 775 | |||
AK152650.1 | - | 5 | 775 | |||
AK152731.1 | - | 5 | 746 | |||
AK152983.1 | - | 5 | 767 | |||
AK153103.1 | - | 5 | 718 | |||
AK159307.1 | - | 5 | 718 | |||
AK159340.1 | - | 5 | 743 | |||
AK159378.1 | - | 5 | 774 | |||
AK159426.1 | - | 5 | 743 | |||
AK159619.1 | - | 5 | 767 | |||
AK159628.1 | - | 5 | 743 | |||
AK159792.1 | - | 5 | 743 | |||
AK159962.1 | - | 5 | 746 | |||
AK160101.1 | - | 5 | 743 | |||
AK160725.1 | - | 5 | 774 | |||
BC004731.1 | - | 5 | 726 | 3583154 | ||
BC010320.1 | - | 5 | 758 | 3599190 | ||
BC021786.1 | - | 5 | 764 | 4458918 | ||
U76253.1 | - | 5 | 755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
