siRNA Details
Gene Symbol: Itsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110275.1 | NP_001103745.1 | 19 | 2455 | ||
NM_010587.2 | NP_034717.2 | 19 | 2455 | |||
GenBank mRNA | AF132478.1 | - | 18 | 2194 | ||
AF132481.1 | - | 18 | 2194 | |||
AK160794.1 | - | 4 | 444 | |||
AY127576.1 | - | 2 | 73 | |||
AY605094.1 | - | 2 | 73 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
