siRNA Details
Gene Symbol: Itsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110275.1 | NP_001103745.1 | 22 | 3033 | ||
NM_010587.2 | NP_034717.2 | 22 | 3033 | |||
GenBank mRNA | AF132478.1 | - | 21 | 2772 | ||
AF132481.1 | - | 21 | 2772 | |||
AK160794.1 | - | 7 | 1022 | |||
AK199389.1 | - | 2 | 228 | |||
BC062938.1 | - | 22 | 3032 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001136096, NM_019227, AF127798, AF132672.
Gene Ontology Categories:
