siRNA Details
Gene Symbol: Jup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010593.2 | NP_034723.1 | 4 | 859 | ||
GenBank mRNA | AK145895.1 | - | 4 | 844 | ||
AK170934.1 | - | 4 | 832 | |||
BC040757.1 | - | 4 | 808 | 4216772 | ||
BC094461.1 | - | 4 | 812 | 4459882 | ||
M90365.1 | - | 2 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
