siRNA Details
Gene Symbol: Krt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010668.2 | NP_034798.2 | 1 | 630 | ||
GenBank mRNA | AK132476.1 | - | 1 | 632 | ||
AK161078.1 | - | 1 | 629 | |||
AK161098.1 | - | 1 | 632 | |||
BC120485.1 | - | 1 | 596 | 40129408 | ||
BC139485.1 | - | 1 | 596 | 8860587 | ||
X74784.1 | - | 1 | 596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
