siRNA Details
Gene Symbol: L1cam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008478.3 | NP_032504.3 | 7 | 825 | ||
GenBank mRNA | AK139407.1 | - | 7 | 787 | ||
BC056988.1 | - | 7 | 750 | 6822217 | ||
X12875.1 | - | 6 | 572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
