siRNA Details
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083188.1 | NP_001076657.1 | 2, 3 | 277 | ||
NM_001199310.1 | NP_001186239.1 | 2, 3 | 285 | |||
NM_010715.2 | NP_034845.2 | 2, 3 | 202 | |||
GenBank mRNA | AK053906.1 | - | 2, 3 | 194 | ||
AK153993.1 | - | 2, 3 | 164 | |||
AK168643.1 | - | 2, 3 | 279 | |||
BC028287.1 | - | 2, 3 | 127 | 4911261 | ||
BC138541.1 | - | 2, 3 | 285 | 8861561 | ||
BC138542.1 | - | 2, 3 | 285 | 8861562 | ||
BY099263.1 | - | 2, 3 | 202 | |||
U04674.1 | - | 1, 2 | 70 | |||
U19604.1 | - | 2, 3 | 315 |

More Information
Other Species Targeted

Gene Ontology Categories:
